Product Details
- SNP ID
-
hCV60771583
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:52300465 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAGGAAGAGGAAAGCAAAGGAGT[C/T]AGGGATGGCTCTTCCTCAGGTGAGA
- Phenotype
-
MIM: 613910
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF480
PubMed Links
Gene Details
- Gene
- ZNF480
- Gene Name
- zinc finger protein 480
There are no transcripts associated with this gene.
- Gene
- ZNF766
- Gene Name
- zinc finger protein 766
There are no transcripts associated with this gene.
View Full Product Details