Product Details

SNP ID
rs28359765
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2270778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGCAACTTTGGGGCCTGTTTA[C/T]CCTGCGCAGCAAATGGCTGCTGGCG
Phenotype
MIM: 608743 MIM: 601579
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C19orf35 PubMed Links
Additional Information
For this assay, SNP(s) [rs112691558] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C19orf35
Gene Name
chromosome 19 open reading frame 35
There are no transcripts associated with this gene.

Gene
JSRP1
Gene Name
junctional sarcoplasmic reticulum protein 1
There are no transcripts associated with this gene.

Gene
OAZ1
Gene Name
ornithine decarboxylase antizyme 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301020.1 Intron NP_001287949.1
NM_004152.3 Intron NP_004143.1

View Full Product Details