Product Details

SNP ID
rs28400331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17361438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCTGGCCTACCCCTGTCTCCA[C/T]CACAGCCTCAGGGTGAATCCAGTTC
Phenotype
MIM: 607647
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLVAP PubMed Links

Gene Details

Gene
PLVAP
Gene Name
plasmalemma vesicle associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031310.2 Intron NP_112600.1

View Full Product Details