Product Details

SNP ID
rs28877890
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29200792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGAAATAAAATTCATTTCCGTA[A/T]ATTGATCTTCTATTCTGCAACCTTA
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
LINC00189 PubMed Links

Gene Details

Gene
LINC00189
Gene Name
long intergenic non-protein coding RNA 189
There are no transcripts associated with this gene.

View Full Product Details