Product Details

SNP ID
rs34188085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39402924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCAGCCGGATAAAACAGTGTTA[G/T]TCTCATACTGGCTTTGGGCAAATCA
Phenotype
MIM: 602615
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TAB1 PubMed Links

Gene Details

Gene
TAB1
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006116.2 Intron NP_006107.1
NM_153497.2 Intron NP_705717.1

View Full Product Details