Product Details

SNP ID
rs28741106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41623033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTAACACAACCACTACCAGCAT[A/T]TTGACATTTTTGTATTTTTCTTTCA
Phenotype
MIM: 614637 MIM: 152690
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DESI1 PubMed Links

Gene Details

Gene
DESI1
Gene Name
desumoylating isopeptidase 1
There are no transcripts associated with this gene.

Gene
XRCC6
Gene Name
X-ray repair cross complementing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288976.1 Intron NP_001275905.1
NM_001288977.1 Intron NP_001275906.1
NM_001288978.1 Intron NP_001275907.1
NM_001469.4 Intron NP_001460.1

View Full Product Details