Product Details
- SNP ID
-
hCV61268770
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:46273919 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTGATGGGCCACGCCATGGCTA[C/T]TGGCCTTGTGCTGATTGGCACTGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTC38
PubMed Links
Gene Details
- Gene
- TTC38
- Gene Name
- tetratricopeptide repeat domain 38
There are no transcripts associated with this gene.
View Full Product Details