Product Details
- SNP ID
-
rs28445694
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50353791 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGATAGAGGCAAGATTTGTACCCC[A/C]GGTAGTGTGACTCAAAAGCCTCTCC
- Phenotype
-
MIM: 610877
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP6R2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111868603] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP6R2
- Gene Name
- protein phosphatase 6 regulatory subunit 2
View Full Product Details