Product Details

SNP ID
rs10783
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72272730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCTCTTCAGCAATGGTGAGGC[G/T]GATACCCTTTCCTCGGGGAAGAGAA
Phenotype
MIM: 300252 MIM: 312760
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
PIN4 PubMed Links

Gene Details

Gene
PIN4
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
There are no transcripts associated with this gene.

Gene
RPS4X
Gene Name
ribosomal protein S4, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007.4 Intron NP_000998.1

View Full Product Details