Product Details

SNP ID
rs28460664
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:16852460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACCACGTACAAGACATTTTTTT[C/T]AATAGTGGACCCACGAGTCTGGTAT
Phenotype
MIM: 300825 MIM: 300677
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RBBP7 PubMed Links

Gene Details

Gene
RBBP7
Gene Name
RB binding protein 7, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198719.1 Intron NP_001185648.1
NM_002893.3 Intron NP_002884.1
Gene
TXLNG
Gene Name
taxilin gamma
There are no transcripts associated with this gene.

View Full Product Details