Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278590.1 | Intron | NP_001265519.1 | |||
NM_001278591.1 | Intron | NP_001265520.1 | |||
NM_001278592.1 | Intron | NP_001265521.1 | |||
NM_006859.3 | Intron | NP_006850.2 | |||
NM_194451.2 | Intron | NP_919433.1 | |||
XM_006713990.3 | Intron | XP_006714053.1 | |||
XM_017007665.1 | Intron | XP_016863154.1 | |||
XM_017007666.1 | Intron | XP_016863155.1 | |||
XM_017007667.1 | Intron | XP_016863156.1 |