Product Details

SNP ID
rs35397046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:109912912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAAACTGTTGGGAGAGGAATC[A/G]TATCTCCATATTTCTTCTTTCAGCC
Phenotype
MIM: 131530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGF PubMed Links

Gene Details

Gene
EGF
Gene Name
epidermal growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178130.2 30 UTR 5 NP_001171601.1
NM_001178131.2 30 UTR 5 NP_001171602.1
NM_001963.5 30 UTR 5 NP_001954.2
XM_005262796.2 30 UTR 5 XP_005262853.1
XM_011531707.1 30 Intron XP_011530009.1
XM_017007845.1 30 UTR 5 XP_016863334.1
XM_017007846.1 30 UTR 5 XP_016863335.1
XM_017007847.1 30 UTR 5 XP_016863336.1
XM_017007848.1 30 UTR 5 XP_016863337.1
XM_017007849.1 30 UTR 5 XP_016863338.1
XM_017007850.1 30 UTR 5 XP_016863339.1
XM_017007851.1 30 UTR 5 XP_016863340.1
XM_017007852.1 30 UTR 5 XP_016863341.1
XM_017007853.1 30 UTR 5 XP_016863342.1
XM_017007854.1 30 UTR 5 XP_016863343.1
XM_017007855.1 30 UTR 5 XP_016863344.1

View Full Product Details