Product Details

SNP ID
rs28450144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:119497530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATATGGCAATTTTTCAATCACA[A/G]TACTGCTAAATGTTATTCAGGCATA
Phenotype
MIM: 603310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC645513 PubMed Links

Gene Details

Gene
LOC645513
Gene Name
septin 7 pseudogene
There are no transcripts associated with this gene.

Gene
PDE5A
Gene Name
phosphodiesterase 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083.3 3685 UTR 3 NP_001074.2
NM_033430.2 3685 UTR 3 NP_236914.2
NM_033437.3 3685 UTR 3 NP_246273.2
XM_017008791.1 3685 Intron XP_016864280.1

View Full Product Details