Product Details
- SNP ID
-
hCV61685170
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:122451297 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAACTAAGGGTCTGATCAGCCCT[C/T]GAAAGGACAAAGCAAGGGCTTTTGA
- Phenotype
-
MIM: 147680
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL2
PubMed Links
Gene Details
- Gene
- IL2
- Gene Name
- interleukin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000586.3 |
|
Intron |
|
|
NP_000577.2 |
View Full Product Details