Product Details

SNP ID
hCV61823065
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.1:103550988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGTCGTATGAAAGGACAAGCTT[C/T]CATTGGACTTCCTAATGAAAAAGCA
Phenotype
MIM: 104660
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AMY2B PubMed Links

Gene Details

Gene
AMY2B
Gene Name
amylase, alpha 2B (pancreatic)
There are no transcripts associated with this gene.

Gene
RNPC3
Gene Name
RNA binding region (RNP1, RRM) containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017619.3 Intron NP_060089.1

View Full Product Details