Product Details

SNP ID
rs35629664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23917934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCATGCGCTTTCTTCTGAGGGT[G/C]CGCTGCTGGCAGTACCGCCAGCTCT
Phenotype
MIM: 604496 MIM: 604174
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
NKIRAS1 PubMed Links

Gene Details

Gene
NKIRAS1
Gene Name
NFKB inhibitor interacting Ras like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020345.3 230 Intron NP_065078.1
XM_005265076.3 230 Intron XP_005265133.1
XM_005265077.4 230 Intron XP_005265134.1
XM_005265078.3 230 Intron XP_005265135.1
Gene
RPL15
Gene Name
ribosomal protein L15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253379.1 230 Silent Mutation GTC,GTG V25V NP_001240308.1
NM_001253380.1 230 Silent Mutation NP_001240309.1
NM_001253382.1 230 Silent Mutation NP_001240311.1
NM_001253383.1 230 Silent Mutation NP_001240312.1
NM_001253384.1 230 Silent Mutation GTC,GTG V25V NP_001240313.1
NM_002948.3 230 Silent Mutation GTC,GTG V25V NP_002939.2

View Full Product Details