Product Details

SNP ID
rs34178602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38166038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGCGGGAGCTCGGGGAGCGCAG[C/T]CCTCATAGGAATTCGTGGGGCGCTT
Phenotype
MIM: 604046
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OXSR1 PubMed Links

Gene Details

Gene
OXSR1
Gene Name
oxidative stress responsive 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005109.2 Intron NP_005100.1
XM_005265638.2 Intron XP_005265695.1
XM_011534331.2 Intron XP_011532633.1
XM_017007601.1 Intron XP_016863090.1
XM_017007602.1 Intron XP_016863091.1

View Full Product Details