Product Details

SNP ID
rs28490287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:62405158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGGGGGGGGGGGGGGCTCAACA[A/G]ATCTATTTTGAGAACTATGATGAAC
Phenotype
MIM: 604667
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CADPS PubMed Links

Gene Details

Gene
CADPS
Gene Name
calcium dependent secretion activator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003716.3 Intron NP_003707.2
NM_183393.2 Intron NP_899630.1
NM_183394.2 Intron NP_899631.1
XM_006713378.2 Intron XP_006713441.2
XM_011534177.1 Intron XP_011532479.1
XM_011534178.1 Intron XP_011532480.1
XM_011534180.1 Intron XP_011532482.1
XM_011534182.1 Intron XP_011532484.1
XM_011534183.1 Intron XP_011532485.1
XM_011534189.1 Intron XP_011532491.1
XM_011534191.2 Intron XP_011532493.1
XM_011534192.1 Intron XP_011532494.1
XM_011534193.1 Intron XP_011532495.1
XM_011534194.2 Intron XP_011532496.1
XM_011534195.1 Intron XP_011532497.1
XM_011534196.1 Intron XP_011532498.1
XM_011534197.2 Intron XP_011532499.1
XM_011534198.2 Intron XP_011532500.1
XM_011534199.2 Intron XP_011532501.1
XM_011534200.2 Intron XP_011532502.1
XM_011534202.2 Intron XP_011532504.1
XM_011534203.1 Intron XP_011532505.1
XM_017007358.1 Intron XP_016862847.1
XM_017007359.1 Intron XP_016862848.1
XM_017007360.1 Intron XP_016862849.1
XM_017007361.1 Intron XP_016862850.1
XM_017007362.1 Intron XP_016862851.1
XM_017007363.1 Intron XP_016862852.1
XM_017007364.1 Intron XP_016862853.1
XM_017007365.1 Intron XP_016862854.1
XM_017007366.1 Intron XP_016862855.1
XM_017007367.1 Intron XP_016862856.1
XM_017007368.1 Intron XP_016862857.1
XM_017007369.1 Intron XP_016862858.1
XM_017007370.1 Intron XP_016862859.1
XM_017007371.1 Intron XP_016862860.1
XM_017007372.1 Intron XP_016862861.1
XM_017007373.1 Intron XP_016862862.1
XM_017007374.1 Intron XP_016862863.1
XM_017007375.1 Intron XP_016862864.1
XM_017007376.1 Intron XP_016862865.1
XM_017007377.1 Intron XP_016862866.1
XM_017007378.1 Intron XP_016862867.1
XM_017007379.1 Intron XP_016862868.1
XM_017007380.1 Intron XP_016862869.1
XM_017007381.1 Intron XP_016862870.1
XM_017007382.1 Intron XP_016862871.1
XM_017007383.1 Intron XP_016862872.1
XM_017007384.1 Intron XP_016862873.1
XM_017007385.1 Intron XP_016862874.1
XM_017007386.1 Intron XP_016862875.1

View Full Product Details