Product Details

SNP ID
rs28372784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9649529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTTCTAGTGTCGGAGTTGGGTG[C/T]AGGCAGGTGCCATGGGCCCGCTTGA
Phenotype
MIM: 611089
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTMR14 PubMed Links

Gene Details

Gene
MTMR14
Gene Name
myotubularin related protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077525.2 64 UTR 5 NP_001070993.1
NM_001077526.2 64 UTR 5 NP_001070994.1
NM_022485.4 64 UTR 5 NP_071930.2
XM_005265400.1 64 UTR 5 XP_005265457.1
XM_011534012.1 64 UTR 5 XP_011532314.1
XM_011534013.1 64 Intron XP_011532315.1
XM_017007034.1 64 UTR 5 XP_016862523.1
XM_017007035.1 64 UTR 5 XP_016862524.1
XM_017007036.1 64 UTR 5 XP_016862525.1
XM_017007037.1 64 UTR 5 XP_016862526.1
XM_017007038.1 64 UTR 5 XP_016862527.1
XM_017007039.1 64 UTR 5 XP_016862528.1
XM_017007040.1 64 UTR 5 XP_016862529.1
XM_017007041.1 64 UTR 5 XP_016862530.1
XM_017007042.1 64 UTR 5 XP_016862531.1
XM_017007043.1 64 UTR 5 XP_016862532.1
XM_017007044.1 64 UTR 5 XP_016862533.1
XM_017007045.1 64 UTR 5 XP_016862534.1

View Full Product Details