Product Details

SNP ID
rs28488962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:188001732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCAGTCTAGAAAGAACCTGGC[A/G]TGTAGCAGGATGCAACATATACTTG
Phenotype
MIM: 614572
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP42 PubMed Links

Gene Details

Gene
ZFP42
Gene Name
ZFP42 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304358.1 Intron NP_001291287.1
NM_174900.4 Intron NP_777560.2
XM_011531604.2 Intron XP_011529906.1
XM_011531605.1 Intron XP_011529907.1
XM_011531606.1 Intron XP_011529908.1
XM_011531607.2 Intron XP_011529909.1

View Full Product Details