Product Details

SNP ID
rs7445128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177701035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTCTCATGAATGGTTTAGTACA[A/G]TCCCCTTGGTGGTGGTCTCGTGATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM153A PubMed Links

Gene Details

Gene
FAM153A
Gene Name
family with sequence similarity 153 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173663.3 Intron NP_775934.3
XM_005265889.3 Intron XP_005265946.1
XM_006714848.3 Intron XP_006714911.1
XM_006714849.3 Intron XP_006714912.1
XM_011534520.2 Intron XP_011532822.1
XM_011534521.2 Intron XP_011532823.1
XM_011534522.2 Intron XP_011532824.1
XM_011534523.2 Intron XP_011532825.1
XM_011534528.2 Intron XP_011532830.1
XM_017009352.1 Intron XP_016864841.1
XM_017009353.1 Intron XP_016864842.1
XM_017009354.1 Intron XP_016864843.1
XM_017009355.1 Intron XP_016864844.1
XM_017009356.1 Intron XP_016864845.1
XM_017009357.1 Intron XP_016864846.1
XM_017009358.1 Intron XP_016864847.1
XM_017009359.1 Intron XP_016864848.1
XM_017009360.1 Intron XP_016864849.1
XM_017009361.1 Intron XP_016864850.1
XM_017009362.1 Intron XP_016864851.1
XM_017009363.1 Intron XP_016864852.1
XM_017009364.1 Intron XP_016864853.1
XM_017009365.1 Intron XP_016864854.1
XM_017009366.1 Intron XP_016864855.1
XM_017009367.1 Intron XP_016864856.1
XM_017009368.1 Intron XP_016864857.1
XM_017009369.1 Intron XP_016864858.1
XM_017009370.1 Intron XP_016864859.1
XM_017009371.1 Intron XP_016864860.1

View Full Product Details