Product Details
- SNP ID
-
rs28364766
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:29067959 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTCTGCTGCTGGACGCAGTGCCC[A/G]GGGCAGAAGTGAGCCTTCTCACCCT
- Phenotype
-
MIM: 607350
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HMBOX1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76644623] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HMBOX1
- Gene Name
- homeobox containing 1
There are no transcripts associated with this gene.
- Gene
- KIF13B
- Gene Name
- kinesin family member 13B
View Full Product Details