Product Details

SNP ID
rs28505577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63910498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTCCCCAGTGTCTATTTTTGCC[A/G]TCTTTAAGTCTATGAGTACCCAATG
Phenotype
MIM: 602058 MIM: 173390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINB10 PubMed Links

Gene Details

Gene
SERPINB10
Gene Name
serpin family B member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005024.2 Intron NP_005015.1
XM_011526027.1 Intron XP_011524329.1
XM_011526028.1 Intron XP_011524330.1
XM_017025793.1 Intron XP_016881282.1
XM_017025794.1 Intron XP_016881283.1
Gene
SERPINB2
Gene Name
serpin family B member 2
There are no transcripts associated with this gene.

View Full Product Details