Product Details

SNP ID
rs28408764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20143999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGTTGGTGGACTTCCTCTCTGA[A/G]AAGAAGGTAATCTCCTATAGAAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4N5 PubMed Links

Gene Details

Gene
OR4N5
Gene Name
olfactory receptor family 4 subfamily N member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004724.1 264 Silent Mutation GAA,GAG E88E NP_001004724.1

View Full Product Details