Product Details

SNP ID
rs35287494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64518470 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTCCTGTATCTCTGGCTATGGAA[C/T]TTTATGTCTTAGAGGTGGCAGGATG
Phenotype
MIM: 616578
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102723809 PubMed Links

Gene Details

Gene
LOC102723809
Gene Name
uncharacterized LOC102723809
There are no transcripts associated with this gene.

Gene
ZBTB1
Gene Name
zinc finger and BTB domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123329.1 Intron NP_001116801.1
NM_014950.2 Intron NP_055765.2
XM_005267410.1 Intron XP_005267467.1
XM_011536566.2 Intron XP_011534868.1
XM_011536568.2 Intron XP_011534870.1
XM_017021094.1 Intron XP_016876583.1
XM_017021095.1 Intron XP_016876584.1

View Full Product Details