Product Details

SNP ID
rs35675176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:102932865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAATCACGGGGGCTCCATGCGGGG[A/G]CAGGACTGGTGGGGGGGGGGGCGGG
Phenotype
MIM: 605799 MIM: 614062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMN PubMed Links

Gene Details

Gene
AMN
Gene Name
amnion associated transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030943.3 6215 Intron NP_112205.2
Gene
CDC42BPB
Gene Name
CDC42 binding protein kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006035.3 6215 UTR 3 NP_006026.3
XM_005268227.1 6215 UTR 3 XP_005268284.1
XM_005268228.1 6215 UTR 3 XP_005268285.1
XM_005268229.1 6215 UTR 3 XP_005268286.1
XM_005268230.4 6215 Intron XP_005268287.1
XM_011537387.1 6215 Intron XP_011535689.1

View Full Product Details