Product Details

SNP ID
rs35409039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34956118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGGGCACACAATTGATAAGGGT[A/G]GGGGTGGCCGTGGTAATGGAGGCCC
Phenotype
MIM: 601002 MIM: 609928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSS PubMed Links

Gene Details

Gene
GSS
Gene Name
glutathione synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000178.3 Intron NP_000169.1
NM_001322494.1 Intron NP_001309423.1
NM_001322495.1 Intron NP_001309424.1
Gene
MYH7B
Gene Name
myosin heavy chain 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020884.4 Intron NP_065935.3
XM_006723840.3 Intron XP_006723903.1
XM_011528941.2 Intron XP_011527243.1
XM_011528947.2 Intron XP_011527249.1
XM_011528948.2 Intron XP_011527250.1
XM_017027986.1 Intron XP_016883475.1
XM_017027987.1 Intron XP_016883476.1

View Full Product Details