Product Details

SNP ID
rs1132734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139561784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCCGGGGGCCGCCGCCACCCGC[A/C]TCCCACCATGGCTCTGAAGAGAATC
Phenotype
MIM: 602962
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBE2D2 PubMed Links

Gene Details

Gene
UBE2D2
Gene Name
ubiquitin conjugating enzyme E2 D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003339.2 619 UTR 5 NP_003330.1
NM_181838.1 619 UTR 5 NP_862821.1
XM_017009820.1 619 UTR 5 XP_016865309.1

View Full Product Details