Product Details
- SNP ID
-
hCV63474416
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:57253769 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGAGAAACAGTGACTGATGTCCAG[G/T]GACAGATTTTTTTTTTTATATTTAA
- Phenotype
-
MIM: 615521
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
R3HDM2
PubMed Links
Gene Details
- Gene
- R3HDM2
- Gene Name
- R3H domain containing 2
There are no transcripts associated with this gene.
- Gene
- STAC3
- Gene Name
- SH3 and cysteine rich domain 3
There are no transcripts associated with this gene.
View Full Product Details