Product Details

SNP ID
rs3782394
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:67653453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGAATCTCTAACATCTGGAAA[C/G]CTTTTAAGATTGCATGTGTTGCTTT
Phenotype
MIM: 603496
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
DYRK2 PubMed Links
Additional Information
For this assay, SNP(s) [rs115700431] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DYRK2
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003583.3 Intron NP_003574.1
NM_006482.2 Intron NP_006473.2
XM_017020032.1 Intron XP_016875521.1

View Full Product Details