Product Details

SNP ID
rs34549734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86665186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCAGCCAGAGGGACCCTGGGGAT[C/T]GGCAGTGGGGGACATAGGAGAAGGG
Phenotype
MIM: 605906 MIM: 606665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDB3 PubMed Links

Gene Details

Gene
LDB3
Gene Name
LIM domain binding 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080114.1 402 Intron NP_001073583.1
NM_001080115.1 402 Intron NP_001073584.1
NM_001080116.1 402 Intron NP_001073585.1
NM_001171610.1 402 Intron NP_001165081.1
NM_001171611.1 402 Intron NP_001165082.1
NM_007078.2 402 Intron NP_009009.1
XM_005269464.4 402 Intron XP_005269521.1
XM_005269466.4 402 Intron XP_005269523.1
XM_005269468.4 402 Intron XP_005269525.1
XM_011539184.2 402 Intron XP_011537486.1
XM_011539185.2 402 UTR 5 XP_011537487.1
XM_011539186.2 402 Intron XP_011537488.1
XM_011539187.2 402 Intron XP_011537489.1
XM_011539188.2 402 Intron XP_011537490.1
XM_011539189.1 402 Intron XP_011537491.1
XM_011539190.1 402 Intron XP_011537492.1
XM_011539191.2 402 Intron XP_011537493.1
XM_011539193.1 402 Intron XP_011537495.1
XM_011539194.1 402 Intron XP_011537496.1
XM_011539195.2 402 Intron XP_011537497.1
XM_017015606.1 402 UTR 5 XP_016871095.1
XM_017015607.1 402 Intron XP_016871096.1
XM_017015608.1 402 UTR 5 XP_016871097.1
XM_017015609.1 402 Intron XP_016871098.1
Gene
OPN4
Gene Name
opsin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030015.2 402 Intron NP_001025186.1
NM_033282.3 402 Intron NP_150598.1
XM_017016955.1 402 Intron XP_016872444.1
XM_017016956.1 402 Intron XP_016872445.1
XM_017016957.1 402 Intron XP_016872446.1

View Full Product Details