Product Details

SNP ID
rs28674759
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:40623380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAAAAGGAGTCTATGAAGAAAG[G/T]GGGACAGAATGAACAGAGCAGAAAT
Phenotype
MIM: 611600
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RIMS3 PubMed Links

Gene Details

Gene
RIMS3
Gene Name
regulating synaptic membrane exocytosis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014747.2 4171 UTR 3 NP_055562.2
XM_011542479.2 4171 UTR 3 XP_011540781.1

View Full Product Details