Product Details

SNP ID
rs28607905
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.1:45550829 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTGCACCGCCCACGTGGCCAGC[A/G]CCACCTGCCTCATTGTGCCCAGGAG
Phenotype
MIM: 103830
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AKR1A1 PubMed Links

Gene Details

Gene
AKR1A1
Gene Name
aldo-keto reductase family 1 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202413.1 47 UTR 5 NP_001189342.1
NM_001202414.1 47 UTR 5 NP_001189343.1
NM_006066.3 47 UTR 5 NP_006057.1
NM_153326.2 47 UTR 5 NP_697021.1
XM_011540491.2 47 Intron XP_011538793.1
XM_011540492.2 47 UTR 5 XP_011538794.1

View Full Product Details