Product Details
- SNP ID
-
rs28492951
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:37539740 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAAAATCTATCCTATCCTTCTCA[C/G]AGGATTGTTGTGAGAATCATCACAG
- Phenotype
-
MIM: 608241
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNIP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200483331] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNIP1
- Gene Name
- Smad nuclear interacting protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024700.3 |
|
Intron |
|
|
NP_078976.2 |
View Full Product Details