Product Details

SNP ID
rs34067094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123725589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAAGACAGAATCCTTCAGGCTC[A/G]TCCCTTCCCAAACCTAAGGGGCTGT
Phenotype
MIM: 603430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF202 PubMed Links

Gene Details

Gene
ZNF202
Gene Name
zinc finger protein 202
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301779.1 2955 UTR 3 NP_001288708.1
NM_001301780.1 2955 UTR 3 NP_001288709.1
NM_001301819.1 2955 UTR 3 NP_001288748.1
NM_003455.3 2955 UTR 3 NP_003446.2
XM_005271659.1 2955 UTR 3 XP_005271716.1
XM_005271660.1 2955 UTR 3 XP_005271717.1
XM_005271661.1 2955 UTR 3 XP_005271718.1
XM_005271664.1 2955 UTR 3 XP_005271721.1
XM_006718901.2 2955 UTR 3 XP_006718964.1
XM_011542972.1 2955 UTR 3 XP_011541274.1
XM_011542973.1 2955 UTR 3 XP_011541275.1
XM_011542975.1 2955 UTR 3 XP_011541277.1
XM_011542976.1 2955 UTR 3 XP_011541278.1
XM_017018268.1 2955 UTR 3 XP_016873757.1

View Full Product Details