Product Details

SNP ID
rs28494624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134372391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCCTGGGCCTCCTTTGGTAAAT[A/G]AAATAGGGTTTTGTTAGGAGGTATT
Phenotype
MIM: 608459 MIM: 179095
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDKL3 PubMed Links

Gene Details

Gene
CDKL3
Gene Name
cyclin dependent kinase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113575.1 Intron NP_001107047.1
NM_001300853.1 Intron NP_001287782.1
NM_016508.3 Intron NP_057592.2
Gene
UBE2B
Gene Name
ubiquitin conjugating enzyme E2 B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003337.3 Intron NP_003328.1

View Full Product Details