Product Details

SNP ID
rs28516593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:154951388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGTGACTGTAGTGGGGGTGGC[A/G]GTGTGTGTGCATGGGTATGGGCTGG
Phenotype
MIM: 608254
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAXIP1 PubMed Links

Gene Details

Gene
PAXIP1
Gene Name
PAX interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007349.3 Intron NP_031375.3
XM_005249539.1 Intron XP_005249596.1
XM_011515982.2 Intron XP_011514284.1
XM_011515983.1 Intron XP_011514285.1
XM_017011881.1 Intron XP_016867370.1
Gene
PAXIP1-AS2
Gene Name
PAXIP1 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details