Product Details

SNP ID
rs28659967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133366892 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACACCAGAAAGTGGAGCCCTTGCC[A/G]TCATAAAGAATGGGCTGTTTCCCAC
Phenotype
MIM: 185630 MIM: 185660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STKLD1 PubMed Links

Gene Details

Gene
STKLD1
Gene Name
serine/threonine kinase like domain containing 1
There are no transcripts associated with this gene.

Gene
SURF2
Gene Name
surfeit 2
There are no transcripts associated with this gene.

Gene
SURF4
Gene Name
surfeit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280788.1 Intron NP_001267717.1
NM_001280789.1 Intron NP_001267718.1
NM_001280790.1 Intron NP_001267719.1
NM_001280791.1 Intron NP_001267720.1
NM_001280792.1 Intron NP_001267721.1
NM_001280793.1 Intron NP_001267722.1
NM_033161.3 Intron NP_149351.1
XM_011518943.1 Intron XP_011517245.1
XM_011518944.2 Intron XP_011517246.1
XM_011518945.2 Intron XP_011517247.1
XM_017015062.1 Intron XP_016870551.1

View Full Product Details