Product Details

SNP ID
rs34519934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130714249 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAAATCCCACGTATATGCCATTT[C/T]CCTCTACGCTCGCTGACCGTTCTGG
Phenotype
MIM: 189980 MIM: 602238
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABL1 PubMed Links

Gene Details

Gene
ABL1
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005157.5 369 Intron NP_005148.2
NM_007313.2 369 UTR 5 NP_009297.2
Gene
EXOSC2
Gene Name
exosome component 2
There are no transcripts associated with this gene.

View Full Product Details