Product Details

SNP ID
rs28510900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:578634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGAGCTGTTTGTTCACATACTA[C/T]GGCTCACTGCAGCCTCGAACTCCTG
Phenotype
MIM: 611780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC143666 PubMed Links

Gene Details

Gene
LOC143666
Gene Name
uncharacterized LOC143666
There are no transcripts associated with this gene.

Gene
PHRF1
Gene Name
PHD and ring finger domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286581.1 Intron NP_001273510.1
NM_001286582.1 Intron NP_001273511.1
NM_001286583.1 Intron NP_001273512.1
NM_020901.3 Intron NP_065952.2
XM_005253025.4 Intron XP_005253082.1
XM_005253027.3 Intron XP_005253084.1
XM_011520236.2 Intron XP_011518538.1
XM_011520237.2 Intron XP_011518539.1

View Full Product Details