Product Details
- SNP ID
-
rs28710288
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70592568 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAAGAAAAAAAAAAAGACTTTTAT[C/T]TATAGATTGTGTTTTTTTTAATGTT
- Phenotype
-
MIM: 601259
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AMBN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs33910648] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AMBN
- Gene Name
- ameloblastin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016519.5 |
|
Intron |
|
|
NP_057603.1 |
View Full Product Details