Product Details

SNP ID
rs61737469
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:3889984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACATTTCTGCATCATCGTCGTC[A/G]TCATCATCATCATCTTTGGCTCTGA
Phenotype
MIM: 601883
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf174 PubMed Links

Gene Details

Gene
C1orf174
Gene Name
chromosome 1 open reading frame 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207356.2 3303 Silent Mutation GAC,GAT D236D NP_997239.2
XM_005244743.3 3303 Silent Mutation GAC,GAT D247D XP_005244800.1
XM_011541323.2 3303 Silent Mutation GAC,GAT D247D XP_011539625.1
Gene
DFFB
Gene Name
DNA fragmentation factor subunit beta
There are no transcripts associated with this gene.

View Full Product Details