Product Details

SNP ID
rs75282796
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:153561364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTATCAACAGACAAAAAAAGTT[C/T]ATTGAATACAAAACTCAAAGGCATC
Phenotype
MIM: 176993
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A2
Gene Name
S100 calcium binding protein A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005978.3 707 UTR 3 NP_005969.1

View Full Product Details