Product Details

SNP ID
rs60632569
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:248387615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTTCAAACTCAGTACCATGAAT[A/G]AAAACAATGAAACCTTGACCAGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2T6 PubMed Links

Gene Details

Gene
OR2T6
Gene Name
olfactory receptor family 2 subfamily T member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005471.1 Intron NP_001005471.1

View Full Product Details