Product Details
- SNP ID
-
rs61749963
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:244843172 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAAACACAACGGCAGCAGCAGCA[A/G]TTGAACAATCTTGAGCATAGAAGTC
- Phenotype
-
MIM: 614698
MIM: 602869
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COX20
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1053697] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COX20
- Gene Name
- COX20, cytochrome c oxidase assembly factor
- Gene
- HNRNPU
- Gene Name
- heterogeneous nuclear ribonucleoprotein U
View Full Product Details