Product Details

SNP ID
rs151242842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29440023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGTATGCAGGCAGGATGAGTG[A/C]AAACACCTCCATGGTGACTGAGTTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR10C1 PubMed Links

Gene Details

Gene
OR10C1
Gene Name
olfactory receptor family 10 subfamily C member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013941.3 85 Missense Mutation GAA,GCA E3A NP_039229.3
Gene
OR5V1
Gene Name
olfactory receptor family 5 subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030876.5 85 Intron NP_110503.3
XM_011514929.2 85 Intron XP_011513231.1
XM_011514930.2 85 Intron XP_011513232.1

View Full Product Details