Product Details

SNP ID
rs77918452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:18783874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCTTTAAAGCAGAAAAACAAGG[A/G]TTTGAACTCAGATAGGCTTTTAAGG
Phenotype
MIM: 607669 MIM: 603674
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL6IP1 PubMed Links

Gene Details

Gene
ARL6IP1
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 1
There are no transcripts associated with this gene.

Gene
RPS15A
Gene Name
ribosomal protein S15a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001019.4 Intron NP_001010.2
NM_001030009.1 Intron NP_001025180.1

View Full Product Details