Product Details

SNP ID
rs61733789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67176952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCAGCACCCCATGGGGCCGCAG[C/G]TGGTCCACATCCTCATAGGACACCT
Phenotype
MIM: 614117 MIM: 602438 MIM: 605235
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXOC3L1 PubMed Links

Gene Details

Gene
EXOC3L1
Gene Name
exocyst complex component 3 like 1
There are no transcripts associated with this gene.

Gene
HSF4
Gene Name
heat shock transcription factor 4
There are no transcripts associated with this gene.

Gene
KIAA0895L
Gene Name
KIAA0895 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040715.1 2050 Missense Mutation CAC,CAG H425Q NP_001035805.1
Gene
NOL3
Gene Name
nucleolar protein 3
There are no transcripts associated with this gene.

View Full Product Details