Product Details

SNP ID
rs17843039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41065355 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTGTATCCAGATGGTAAAAATCA[C/T]TATTGTCACCTTGTGAAAAAAAATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP2-3 PubMed Links

Gene Details

Gene
KRTAP2-3
Gene Name
keratin associated protein 2-3
There are no transcripts associated with this gene.

Gene
KRTAP2-4
Gene Name
keratin associated protein 2-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033184.3 525 UTR 3 NP_149440.1

View Full Product Details