Product Details

SNP ID
rs138208354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41065430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCAGATAGATGTTTAGGCTTCA[A/G]TGTATTGCTCTGTGGGGGCATTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP2-3 PubMed Links

Gene Details

Gene
KRTAP2-3
Gene Name
keratin associated protein 2-3
There are no transcripts associated with this gene.

Gene
KRTAP2-4
Gene Name
keratin associated protein 2-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033184.3 450 UTR 3 NP_149440.1

View Full Product Details